missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoro-2-cyanopyridine, 97%

Catalog No. AC451430050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451430050 5 g
AC451430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451430050 Supplier Thermo Scientific Chemicals Supplier No. 451430050

CAS: 950670-18-5 | C6H2BrFN2 | 201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 950670-18-5
Molecular Formula C6H2BrFN2
Molecular Weight (g/mol) 201
InChI Key OLOSREGULYFXDD-UHFFFAOYSA-N
Synonym 3-bromo-5-fluoropicolinonitrile, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, abbypharma ap-18-5887, 3-aromo-5-fluoropicolinonitrile
PubChem CID 46941477
IUPAC Name 3-bromo-5-fluoropyridine-2-carbonitrile
SMILES C1=C(C=NC(=C1Br)C#N)F

Specifications

Melting Point 88.0°C to 91.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 201
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluoro-2-cyanopyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

RUO – Research Use Only