missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chlorosalicylaldehyde 96.0+%, TCI America™

Catalog No. p-7119373
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19485G 5 g
B194825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19485G Supplier TCI America Supplier No. B19485G
Only null left

Chemical Identifiers

CAS 19652-32-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00051690
InChI Key KNOYZLVIXXBBIB-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
PubChem CID 519676
IUPAC Name 3-bromo-5-chloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Cl

Specifications

Melting Point 88°C
Quantity 5 g
Formula Weight 235.46
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chlorosalicylaldehyde