missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloropyridine 98.0+%, TCI America™

Catalog No. B36081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36081G 1 g
B36085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36081G Supplier TCI America Supplier No. B36081G

Chemical Identifiers

CAS 73583-39-8
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD04114221
InChI Key BELDOPUBSLPBCQ-UHFFFAOYSA-N
Synonym 3-chloro-5-bromopyridine, 3-bromo-5-chloro-pyridine, pyridine, 3-bromo-5-chloro, 5-bromo-3-chloropyridine, pubchem7700, 3-brom-5-chlorpyridin, acmc-209orf, #, ksc377a4j, 3-bromo-5-chloropyridine
PubChem CID 606256
IUPAC Name 3-bromo-5-chloropyridine
SMILES C1=C(C=NC=C1Br)Cl

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chloropyridine