missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloropyridine, 97%

Catalog No. AC437400250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437400250 25 g
AC437400050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437400250 Supplier Thermo Scientific Chemicals Supplier No. 437400250

CAS: 73583-39-8 | C5H3BrClN | 192.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73583-39-8
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
InChI Key BELDOPUBSLPBCQ-UHFFFAOYSA-N
Synonym 3-chloro-5-bromopyridine, 3-bromo-5-chloro-pyridine, pyridine, 3-bromo-5-chloro, 5-bromo-3-chloropyridine, pubchem7700, 3-brom-5-chlorpyridin, acmc-209orf, #, ksc377a4j, 3-bromo-5-chloropyridine
PubChem CID 606256
IUPAC Name 3-bromo-5-chloropyridine
SMILES C1=C(C=NC=C1Br)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 78.7°C to 80.1°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 192.44
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only