missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-chloro-2-methoxypyridine 98.0+%, TCI America™
$184.54 - $546.17
Chemical Identifiers
| CAS | 102830-75-1 |
|---|---|
| Molecular Formula | C6H5BrClNO |
| Molecular Weight (g/mol) | 222.466 |
| MDL Number | MFCD08059322 |
| InChI Key | AYEVELLBDURPJW-UHFFFAOYSA-N |
| PubChem CID | 13567751 |
| IUPAC Name | 3-bromo-5-chloro-2-methoxypyridine |
| SMILES | COC1=C(C=C(C=N1)Cl)Br |
Chemical Identifiers
| 102830-75-1 | |
| 222.466 | |
| AYEVELLBDURPJW-UHFFFAOYSA-N | |
| 3-bromo-5-chloro-2-methoxypyridine |
| C6H5BrClNO | |
| MFCD08059322 | |
| 13567751 | |
| COC1=C(C=C(C=N1)Cl)Br |
Specifications
| 102830-75-1 | |
| White | |
| MFCD08059322 | |
| AYEVELLBDURPJW-UHFFFAOYSA-N | |
| 3-bromo-5-chloro-2-methoxypyridine | |
| 13567751 | |
| ≥98.0% (GC) | |
| 3-Bromo-5-chloro-2-methoxypyridine |
| 52°C | |
| C6H5BrClNO | |
| 1 g | |
| COC1=C(C=C(C=N1)Cl)Br | |
| 222.466 | |
| 222.47 | |
| Crystalline Powder |
Safety and Handling
TSCA : No