missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloro-2-methoxypyridine 98.0+%, TCI America™

Catalog No. B51655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51655G 5 g
B51651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51655G Supplier TCI America Supplier No. B51655G
Only null left

Chemical Identifiers

CAS 102830-75-1
Molecular Formula C6H5BrClNO
Molecular Weight (g/mol) 222.466
MDL Number MFCD08059322
InChI Key AYEVELLBDURPJW-UHFFFAOYSA-N
PubChem CID 13567751
IUPAC Name 3-bromo-5-chloro-2-methoxypyridine
SMILES COC1=C(C=C(C=N1)Cl)Br

Specifications

Melting Point 52°C
Color White
Quantity 5 g
Formula Weight 222.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chloro-2-methoxypyridine