missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%

Catalog No. AAA1344906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1344906 5 g
AAA1344914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1344906 Supplier Thermo Scientific Chemicals Supplier No. A1344906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19652-32-5 | C7H4BrClO2 | 235.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19652-32-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00051690
InChI Key KNOYZLVIXXBBIB-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
PubChem CID 519676
IUPAC Name 3-bromo-5-chloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Cl

Specifications

Melting Point 84°C to 88°C
Quantity 5 g
Beilstein 2362159
Sensitivity Air Sensitive
Formula Weight 235.47
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-chloro-2-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only