missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-pyridinecarboxaldehyde, 97%

Catalog No. AC398080050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398080050 5 g Glass bottle
AC398080010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398080050 Supplier Thermo Scientific Chemicals Supplier No. 398080050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70201-43-3 | C6H4BrNO | 186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70201-43-3
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186
InChI Key NOBDKWLIAQKADB-UHFFFAOYSA-N
Synonym 3-bromoisonicotinaldehyde, 3-bromo-4-pyridinecarboxaldehyde, 3-bromopyridine-4-carboxaldehyde, 3-bromo-4-formylpyridine, 3-bromo-4-pyridinecarbaldehyde, 4-pyridinecarboxaldehyde, 3-bromo, 3-bromo-4-pyridincarbaldehyde, bromoisonicotinaldehyde, pubchem5063, acmc-1blzq
PubChem CID 2762997
IUPAC Name 3-bromopyridine-4-carbaldehyde
SMILES C1=CN=CC(=C1C=O)Br

Specifications

Melting Point 80°C to 83°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 186
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-pyridinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only