missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitroindole, 97%

CAS: 126807-08-7 | C8H5BrN2O2 | 241.044 g/mol

Supplier:  Thermo Scientific Chemicals H6347803

 View more versions of this product

Catalog No. AAH6347803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-4-nitroindole
C8H5BrN2O2
3-bromo-4-nitroindole, 1h-indole,3-bromo-4-nitro, acmc-20dlf4, 1h-indole, 3-bromo-4-nitro
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)Br
241.044
241.04
Solid
126807-08-7
1 g
VYQRPDVMVMLVHA-UHFFFAOYSA-N
3-bromo-4-nitro-1H-indole
14707933
97%

Chemical Identifiers

126807-08-7
241.044
3-bromo-4-nitroindole, 1h-indole,3-bromo-4-nitro, acmc-20dlf4, 1h-indole, 3-bromo-4-nitro
3-bromo-4-nitro-1H-indole
C8H5BrN2O2
VYQRPDVMVMLVHA-UHFFFAOYSA-N
14707933
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only