missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitroindole, 97%

Catalog No. AAH63478MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63478MD 250 mg
AAH6347803 1 g
AAH6347806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63478MD Supplier Thermo Scientific Chemicals Supplier No. H63478MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 126807-08-7 | C8H5BrN2O2 | 241.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126807-08-7
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
InChI Key VYQRPDVMVMLVHA-UHFFFAOYSA-N
Synonym 3-bromo-4-nitroindole, 1h-indole,3-bromo-4-nitro, acmc-20dlf4, 1h-indole, 3-bromo-4-nitro
PubChem CID 14707933
IUPAC Name 3-bromo-4-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)Br

Specifications

Quantity 250 mg
Formula Weight 241.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-4-nitroindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only