missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylpyridine 98.0+%, TCI America™

Catalog No. B28311G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28311G 1 g
B28315G 5 g
B283125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B28311G Supplier TCI America Supplier No. B28311G

Chemical Identifiers

CAS 3430-22-6
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
InChI Key GSQZOLXWFQQJHJ-UHFFFAOYSA-N
Synonym 3-bromo-4-picoline, 4-methyl-3-bromopyridine, pyridine, 3-bromo-4-methyl, 3-bromo-4-methyl-pyridine, 3-brom-4-methylpyridin, zlchem 494, pubchem3994, 3-bromo4-methylpyridine, 3-bromo-4-methylpyridin
PubChem CID 817630
IUPAC Name 3-bromo-4-methylpyridine
SMILES CC1=C(C=NC=C1)Br

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methylpyridine