missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylpyridine, 98%

Catalog No. AAL1962206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962206 5 g
AAL1962214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1962206 Supplier Thermo Scientific Chemicals Supplier No. L1962206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3430-22-6 | C6H6BrN | 172.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3430-22-6
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00082592
InChI Key GSQZOLXWFQQJHJ-UHFFFAOYSA-N
Synonym 3-bromo-4-picoline, 4-methyl-3-bromopyridine, pyridine, 3-bromo-4-methyl, 3-bromo-4-methyl-pyridine, 3-brom-4-methylpyridin, zlchem 494, pubchem3994, 3-bromo4-methylpyridine, 3-bromo-4-methylpyridin
PubChem CID 817630
IUPAC Name 3-bromo-4-methylpyridine
SMILES CC1=C(C=NC=C1)Br

Specifications

Density 1.549
Boiling Point 199°C to 200°C
Flash Point 79°C (174°F)
Refractive Index 1.562
Quantity 5 g
Beilstein 878354
Formula Weight 172.03
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methylpyridine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only