missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Bromo-4'-methylacetophenone 98.0+%, TCI America™
$50.33 - $154.71
Chemical Identifiers
| CAS | 40180-80-1 |
|---|---|
| Molecular Formula | C9H9BrO |
| Molecular Weight (g/mol) | 213.074 |
| MDL Number | MFCD02683854 |
| InChI Key | PDJLFESXPVLVMR-UHFFFAOYSA-N |
| PubChem CID | 3309401 |
| IUPAC Name | 1-(3-bromo-4-methylphenyl)ethanone |
| SMILES | CC1=C(C=C(C=C1)C(=O)C)Br |
Chemical Identifiers
| 40180-80-1 | |
| 213.074 | |
| PDJLFESXPVLVMR-UHFFFAOYSA-N | |
| 1-(3-bromo-4-methylphenyl)ethanone |
| C9H9BrO | |
| MFCD02683854 | |
| 3309401 | |
| CC1=C(C=C(C=C1)C(=O)C)Br |
Specifications
| 40180-80-1 | |
| White-Yellow | |
| C9H9BrO | |
| 1 g | |
| CC1=C(C=C(C=C1)C(=O)C)Br | |
| 213.074 | |
| 213.07 | |
| Crystalline Powder |
| 44°C | |
| 118°C | |
| MFCD02683854 | |
| PDJLFESXPVLVMR-UHFFFAOYSA-N | |
| 1-(3-bromo-4-methylphenyl)ethanone | |
| 3309401 | |
| ≥98.0% (GC) | |
| 3′-Bromo-4′-methylacetophenone |
Safety and Handling
TSCA : No