missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-4'-methylacetophenone 98.0+%, TCI America™

Catalog No. B47585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47585G 5 g
B47581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47585G Supplier TCI America Supplier No. B47585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40180-80-1
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD02683854
InChI Key PDJLFESXPVLVMR-UHFFFAOYSA-N
PubChem CID 3309401
IUPAC Name 1-(3-bromo-4-methylphenyl)ethanone
SMILES CC1=C(C=C(C=C1)C(=O)C)Br

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-4′-methylacetophenone