missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. p-7035806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5401506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5401506 Supplier Thermo Scientific Chemicals Supplier No. H5401506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23094-96-4 | Molecular Formula: C7H6BrClO3S | Molecular Weight: 285.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23094-96-4
Molecular Formula C7H6BrClO3S
Molecular Weight (g/mol) 285.54
MDL Number MFCD03989643
InChI Key LTVLATJRJIBHDR-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybenzene-1-sulfonyl chloride, 3-bromo-4-methoxy-benzenesulfonyl chloride, 3-bromo-4-methoxyphenylsulfonyl chloride, 3-bromo-4-methoxyphenyl chlorosulfone, 3-bromo4-methoxyphenylsulfonyl chloride, 3-bromo-4-methoxy-benzene-sulfonyl chloride, benzenesulfonylchloride, 3-bromo-4-methoxy, benzenesulfonyl chloride, 3-bromo-4-methoxy, 3-bromo-4-methoxybenzenesulfonyl chloride
PubChem CID 3863747
IUPAC Name 3-bromo-4-methoxybenzenesulfonyl chloride
SMILES COC1=C(Br)C=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 285.54
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methoxybenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only