missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-iodopyridine, 97%

Catalog No. p-7032652
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34203MD 250 mg
AAH3420303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34203MD Supplier Thermo Scientific Chemicals Supplier No. H34203MD

CAS: 89167-19-1 | C5H3BrIN | 283.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89167-19-1
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD11110263
InChI Key LKDNPXCBLVOBRP-UHFFFAOYSA-N
Synonym pyridine, 3-bromo-4-iodo, acmc-209qyv
PubChem CID 52911177
IUPAC Name 3-bromo-4-iodopyridine
SMILES C1=CN=CC(=C1I)Br

Specifications

Melting Point 111°C to 115°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 283.89
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only