Learn More
3-Bromo-4-hydroxybenzaldehyde, 97+%
CAS: 2973-78-6 | C7H5BrO2 | 201.02 g/mol
$82.08 - $273.62
Chemical Identifiers
| CAS | 2973-78-6 |
|---|---|
| Molecular Formula | C7H5BrO2 |
| Molecular Weight (g/mol) | 201.02 |
| MDL Number | MFCD00017348 |
| InChI Key | UOTMHAOCAJROQF-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 3-bromo-4-hydroxy, 3-bromo-4-hydroxy-benzaldehyde, 4-hydroxy-3-bromobenzaldehyde, pubchem3778, acmc-209hav, 2-bromo-4-formylphenol, asischem n48923, 3-bromo-4-hydroxybenzaldehye, ksc490m5b, akos akm00117 |
| PubChem CID | 76308 |
| IUPAC Name | 3-bromo-4-hydroxybenzaldehyde |
| SMILES | OC1=CC=C(C=O)C=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2472906
|
Thermo Scientific Chemicals
B2472906 |
5 g |
Each for $82.08
|
|
|||||
|
AAB2472914
|
Thermo Scientific Chemicals
B2472914 |
25 g |
Each for $273.62
|
|
|||||
Description
3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.
Solubility
Soluble in water 1.33 g/L.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Chemical Identifiers
| 2973-78-6 | |
| 201.02 | |
| UOTMHAOCAJROQF-UHFFFAOYSA-N | |
| 76308 | |
| OC1=CC=C(C=O)C=C1Br |
| C7H5BrO2 | |
| MFCD00017348 | |
| benzaldehyde, 3-bromo-4-hydroxy, 3-bromo-4-hydroxy-benzaldehyde, 4-hydroxy-3-bromobenzaldehyde, pubchem3778, acmc-209hav, 2-bromo-4-formylphenol, asischem n48923, 3-bromo-4-hydroxybenzaldehye, ksc490m5b, akos akm00117 | |
| 3-bromo-4-hydroxybenzaldehyde |
Specifications
| 2973-78-6 | |
| C7H5BrO2 | |
| 5 g | |
| Air Sensitive | |
| Soluble in water 1.33g/L. | |
| OC1=CC=C(C=O)C=C1Br | |
| 201.02 | |
| 201.02 | |
| 3-Bromo-4-hydroxybenzaldehyde |
| 125°C to 128°C | |
| MFCD00017348 | |
| 2205318 | |
| benzaldehyde, 3-bromo-4-hydroxy, 3-bromo-4-hydroxy-benzaldehyde, 4-hydroxy-3-bromobenzaldehyde, pubchem3778, acmc-209hav, 2-bromo-4-formylphenol, asischem n48923, 3-bromo-4-hydroxybenzaldehye, ksc490m5b, akos akm00117 | |
| UOTMHAOCAJROQF-UHFFFAOYSA-N | |
| 3-bromo-4-hydroxybenzaldehyde | |
| 76308 | |
| ≥97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only