missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzaldehyde, 97+%

Catalog No. AAB2472906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2472906 5 g
AAB2472914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2472906 Supplier Thermo Scientific Chemicals Supplier No. B2472906

CAS: 2973-78-6 | C7H5BrO2 | 201.02 g/mol

3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.

Solubility
Soluble in water 1.33 g/L.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 2973-78-6
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00017348
InChI Key UOTMHAOCAJROQF-UHFFFAOYSA-N
Synonym benzaldehyde, 3-bromo-4-hydroxy, 3-bromo-4-hydroxy-benzaldehyde, 4-hydroxy-3-bromobenzaldehyde, pubchem3778, acmc-209hav, 2-bromo-4-formylphenol, asischem n48923, 3-bromo-4-hydroxybenzaldehye, ksc490m5b, akos akm00117
PubChem CID 76308
IUPAC Name 3-bromo-4-hydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C=C1Br

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Beilstein 2205318
Sensitivity Air Sensitive
Solubility Information Soluble in water 1.33g/L.
Formula Weight 201.02
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-4-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only