missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-4'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. B51581G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B51581G Supplier TCI America Supplier No. B51581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1836-06-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD05663950
InChI Key LEXAYGVHWYOOAB-UHFFFAOYSA-N
PubChem CID 608094
IUPAC Name 1-(3-bromo-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)Br

Specifications

Melting Point 113°C
Quantity 1 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-4′-hydroxyacetophenone