missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluoro-5-iodobenzaldehyde, 97%, Thermo Scientific™

Catalog No. AAH3344103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3344103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3344103 Supplier Thermo Scientific Chemicals Supplier No. H3344103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1356113-37-5
Molecular Formula C7H3BrFIO
Molecular Weight (g/mol) 328.91
MDL Number MFCD20486165
InChI Key VZRIOLAMLBZGDB-UHFFFAOYSA-N
PubChem CID 95567744
IUPAC Name 3-bromo-4-fluoro-5-iodobenzaldehyde
SMILES FC1=C(Br)C=C(C=O)C=C1I

Specifications

Quantity 1 g
Formula Weight 328.9
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-fluoro-5-iodobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen