missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-ethoxybenzaldehyde 97.0+%, TCI America™

Catalog No. b50791g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50791G 1 g
B50795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50791G Supplier TCI America Supplier No. B50791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108373-05-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00672941
InChI Key TZUUPGZANQRCHD-UHFFFAOYSA-N
PubChem CID 1494336
IUPAC Name 3-bromo-4-ethoxybenzaldehyde
SMILES CCOC1=C(C=C(C=C1)C=O)Br

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-ethoxybenzaldehyde