missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chloropyridine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 36953-42-1
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD00234008
InChI Key QADXKWUCCGPQNR-UHFFFAOYSA-N
Synonym pyridine, 3-bromo-4-chloro, 3-bromo-4-chloro-pyridine, 4-chloro-3-bromopyridine, acmc-1ai1d, 3-bromo-4-chloropyridine,, ksc222g5j, pyridine,3-bromo-4-chloro-,hydrochloride 1:1
PubChem CID 817696
IUPAC Name 3-bromo-4-chloropyridine
SMILES C1=CN=CC(=C1Cl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC379640010
SDS View Documents
Thermo Scientific Chemicals
379640010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC379640050
SDS View Documents
Thermo Scientific Chemicals
379640050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

36953-42-1
192.44
QADXKWUCCGPQNR-UHFFFAOYSA-N
817696
C1=CN=CC(=C1Cl)Br
C5H3BrClN
MFCD00234008
pyridine, 3-bromo-4-chloro, 3-bromo-4-chloro-pyridine, 4-chloro-3-bromopyridine, acmc-1ai1d, 3-bromo-4-chloropyridine,, ksc222g5j, pyridine,3-bromo-4-chloro-,hydrochloride 1:1
3-bromo-4-chloropyridine

Specifications

36953-42-1
100.0
97.5% min. (GC)
C5H3BrClN
1g
QADXKWUCCGPQNR-UHFFFAOYSA-N
3-bromo-4-chloropyridine
817696
98%
97.5
Authentic
Glass bottle
MFCD00234008
pyridine, 3-bromo-4-chloro, 3-bromo-4-chloro-pyridine, 4-chloro-3-bromopyridine, acmc-1ai1d, 3-bromo-4-chloropyridine,, ksc222g5j, pyridine,3-bromo-4-chloro-,hydrochloride 1:1
C1=CN=CC(=C1Cl)Br
192.44
192.44
3-Bromo-4-chloropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only