missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7119972
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31305G 5 g
B313025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31305G Supplier TCI America Supplier No. B31305G

Chemical Identifiers

CAS 6948-30-7
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00003346
InChI Key ICVODPFGWCUVJC-UHFFFAOYSA-N
Synonym 5-bromoveratraldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 3,4-dimethoxy-5-bromobenzaldehyde, benzaldehyde,3-bromo-4,5-dimethoxy, benzaldehyde, 3-bromo-4,5-dimethoxy, 5-bromoveratric aldehyde, acmc-209ml7, cambridge id 7187634, 5-bromveratrumaldehyde
PubChem CID 81379
IUPAC Name 3-bromo-4,5-dimethoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)Br)OC

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4,5-dimethoxybenzaldehyde