missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4',4″-dimethyltriphenylamine 97.0+%, TCI America™

Catalog No. B50191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B50191G Supplier TCI America Supplier No. B50191G
Only null left

Chemical Identifiers

CAS 845526-91-2
Molecular Formula C20H18BrN
Molecular Weight (g/mol) 352.28
MDL Number MFCD15144691
InChI Key LBLFZUNRAGUAFR-UHFFFAOYSA-N
Synonym 3-Bromo-N,N-di-p-tolylaniline
PubChem CID 51358452
IUPAC Name N-(3-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC(Br)=CC=C1

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 1 g
Formula Weight 352.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4′,4′′-dimethyltriphenylamine