missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals

Catalog No. AC434460010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434460010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434460010 Supplier Thermo Scientific Chemicals Supplier No. 434460010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1514-82-5
Molecular Formula C3H2BrF3
Molecular Weight (g/mol) 174.95
MDL Number MFCD00077469
InChI Key QKBKGNDTLQFSEU-UHFFFAOYSA-N
Synonym 2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
PubChem CID 272696
IUPAC Name 2-bromo-3,3,3-trifluoroprop-1-ene
SMILES FC(F)(F)C(Br)=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.6500g/mL
Boiling Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3520 to 1.3550 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.65
Formula Weight 174.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3, 3, 3-trifluoro-1-propene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only