missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-3,3-difluoropropene 95.0+%, TCI America™
$385.86 - $1733.57
Chemical Identifiers
| CAS | 420-90-6 |
|---|---|
| Molecular Formula | C3H3BrF2 |
| Molecular Weight (g/mol) | 156.958 |
| MDL Number | MFCD00042185 |
| InChI Key | GDDNTTHUKVNJRA-UHFFFAOYSA-N |
| Synonym | 3,3-Difluoro-3-bromopropene |
| PubChem CID | 555085 |
| IUPAC Name | 3-bromo-3,3-difluoroprop-1-ene |
| SMILES | C=CC(F)(F)Br |
Chemical Identifiers
| 420-90-6 | |
| 156.958 | |
| GDDNTTHUKVNJRA-UHFFFAOYSA-N | |
| 555085 | |
| C=CC(F)(F)Br |
| C3H3BrF2 | |
| MFCD00042185 | |
| 3,3-Difluoro-3-bromopropene | |
| 3-bromo-3,3-difluoroprop-1-ene |
Specifications
| 420-90-6 | |
| 42°C | |
| MFCD00042185 | |
| 3,3-Difluoro-3-bromopropene | |
| C=CC(F)(F)Br | |
| 156.958 | |
| 156.96 | |
| Liquid |
| Yellow | |
| C3H3BrF2 | |
| 1 g | |
| GDDNTTHUKVNJRA-UHFFFAOYSA-N | |
| 3-bromo-3,3-difluoroprop-1-ene | |
| 555085 | |
| ≥95.0% (GC) | |
| 3-Bromo-3,3-difluoropropene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator