missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-3,3-difluoropropene 95.0+%, TCI America™

Catalog No. B23331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23331G 1 g
B23335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23331G Supplier TCI America Supplier No. B23331G

Chemical Identifiers

CAS 420-90-6
Molecular Formula C3H3BrF2
Molecular Weight (g/mol) 156.958
MDL Number MFCD00042185
InChI Key GDDNTTHUKVNJRA-UHFFFAOYSA-N
Synonym 3,3-Difluoro-3-bromopropene
PubChem CID 555085
IUPAC Name 3-bromo-3,3-difluoroprop-1-ene
SMILES C=CC(F)(F)Br

Specifications

Color Yellow
Boiling Point 42°C
Quantity 1 g
Formula Weight 156.96
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-3,3-difluoropropene
TSCA No
Recommended Storage Refrigerator