missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitropyridine 98.0+%, TCI America™

Catalog No. p-7123690
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46901G 1 g
B46905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46901G Supplier TCI America Supplier No. B46901G
Only null left

Chemical Identifiers

CAS 54231-33-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00955614
InChI Key WFNISJZUJCKTLT-UHFFFAOYSA-N
Synonym 2-nitro-3-bromopyridine, pyridine, 3-bromo-2-nitro, 3-bromo-2-nitro-pyridine, pubchem4502, pubchem10100, acmc-1ay9l, #, ksc494a7n, 3-bromo-2-nitriopyridine
PubChem CID 594044
IUPAC Name 3-bromo-2-nitropyridine
SMILES [O-][N+](=O)C1=NC=CC=C1Br

Specifications

Melting Point 104°C
Color Yellow
Quantity 1 g
Formula Weight 203.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-nitropyridine