missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitropyridine, 98+%

Catalog No. aal1952303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1952303 1 g
AAL1952306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1952303 Supplier Thermo Scientific Chemicals Supplier No. L1952303

CAS: 54231-33-3 | C5H3BrN2O2 | 203.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54231-33-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00955614
InChI Key WFNISJZUJCKTLT-UHFFFAOYSA-N
Synonym 2-nitro-3-bromopyridine, pyridine, 3-bromo-2-nitro, 3-bromo-2-nitro-pyridine, pubchem4502, pubchem10100, acmc-1ay9l, #, ksc494a7n, 3-bromo-2-nitriopyridine
PubChem CID 594044
IUPAC Name 3-bromo-2-nitropyridine
SMILES [O-][N+](=O)C1=NC=CC=C1Br

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 1637071
Formula Weight 203
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2-nitropyridine
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only