missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog No. p-7053609
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6116906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6116906 Supplier Thermo Scientific Chemicals Supplier No. H6116906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7138-15-0
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD08063248
InChI Key GLKHLBAXHLAQBJ-UHFFFAOYSA-N
Synonym 3-bromo-2-nitrobenzenamine, 2-amino-6-bromonitrobenzene, 3-bromo-2-nitrophenylamine, 3-bromo-2-nitro-phenylamine, pubchem3139, acmc-209oig, 3-bromo-2-nitro-aniline, ksc376s4f, 3-amino-2-nitrobromobenzene
PubChem CID 21852072
IUPAC Name 3-bromo-2-nitroaniline
SMILES C1=CC(=C(C(=C1)Br)[N+](=O)[O-])N

Specifications

Color Red
Quantity 5 g
UN Number UN2811
Formula Weight 217.02
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 3-Bromo-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only