missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-2-furoyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 58777-57-4
Molecular Formula C5H2BrClO2
Molecular Weight (g/mol) 209.423
MDL Number MFCD11109330
InChI Key WTZDPJVRZQGTET-UHFFFAOYSA-N
Synonym 3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci
PubChem CID 21864576
IUPAC Name 3-bromofuran-2-carbonyl chloride
SMILES C1=COC(=C1Br)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC67202DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC67202DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67202DE
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC67202DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

58777-57-4
209.423
WTZDPJVRZQGTET-UHFFFAOYSA-N
21864576
C1=COC(=C1Br)C(=O)Cl
C5H2BrClO2
MFCD11109330
3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci
3-bromofuran-2-carbonyl chloride

Specifications

58777-57-4
100.0
Amber Glass Bottle
MFCD11109330
3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci
C1=COC(=C1Br)C(=O)Cl
209.423
209.43
3-Bromo-2-furoyl chloride
96.5
97%
C5H2BrClO2
1g
WTZDPJVRZQGTET-UHFFFAOYSA-N
3-bromofuran-2-carbonyl chloride
21864576
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'