missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-furoic acid, 97%, Thermo Scientific™

Catalog No. p-3629697
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67201CB 250mg Amber Glass Bottle
CC67201DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67201CB Supplier Thermo Scientific Chemicals Supplier No. CC67201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14903-90-3
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00092761
InChI Key UZBGSJZFBUOJNE-UHFFFAOYSA-N
Synonym 3-bromo-2-furoic acid, furancarboxylic acid, bromo, 3-bromo-2-furancarboxylic acid, 3-bromo-furan-2-carboxylic acid, 3-bromofuran-2-carboxylicacid, bromofuroic acid, 3-bomo-2-furoic acid, 2-furancarboxylic acid,3-bromo, 3-bromanylfuran-2-carboxylic acid, 2-furancarboxylic acid, 3-bromo
PubChem CID 605479
IUPAC Name 3-bromofuran-2-carboxylic acid
SMILES C1=COC(=C1Br)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 190.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-furoic acid