Learn More
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine, 97%
CAS: 1031929-01-7 | C6H2BrF4N | 243.99 g/mol
$63.03 - $605.49
Chemical Identifiers
| CAS | 1031929-01-7 |
|---|---|
| Molecular Formula | C6H2BrF4N |
| Molecular Weight (g/mol) | 243.99 |
| MDL Number | MFCD08061304 |
| InChI Key | HKACHYLUXHBYMO-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-bromo-5-trifluoromethylpyridine, pyridine, 3-bromo-2-fluoro-5-trifluoromethyl, 3-bromo-2-fluoro-5-trifluoromethyl ryridine, pubchem15149, pyridine,3-bromo-2-fluoro-5-trifluoromethyl |
| PubChem CID | 23053237 |
| IUPAC Name | 3-bromo-2-fluoro-5-(trifluoromethyl)pyridine |
| SMILES | FC1=C(Br)C=C(C=N1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH64748MD
|
Thermo Scientific Chemicals
H64748MD |
250 mg |
Each for $63.03
|
|
|||||
|
AAH6474803
|
Thermo Scientific Chemicals
H6474803 |
1 g |
Each for $169.30
|
|
|||||
|
AAH6474806
|
Thermo Scientific Chemicals
H6474806 |
5 g |
Each for $605.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1031929-01-7 | |
| 243.99 | |
| HKACHYLUXHBYMO-UHFFFAOYSA-N | |
| 23053237 | |
| FC1=C(Br)C=C(C=N1)C(F)(F)F |
| C6H2BrF4N | |
| MFCD08061304 | |
| 3-bromo-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-bromo-5-trifluoromethylpyridine, pyridine, 3-bromo-2-fluoro-5-trifluoromethyl, 3-bromo-2-fluoro-5-trifluoromethyl ryridine, pubchem15149, pyridine,3-bromo-2-fluoro-5-trifluoromethyl | |
| 3-bromo-2-fluoro-5-(trifluoromethyl)pyridine |
Specifications
| 1031929-01-7 | |
| 30°C (86°F) | |
| 1.46 | |
| 250 mg | |
| 3-bromo-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-bromo-5-trifluoromethylpyridine, pyridine, 3-bromo-2-fluoro-5-trifluoromethyl, 3-bromo-2-fluoro-5-trifluoromethyl ryridine, pubchem15149, pyridine,3-bromo-2-fluoro-5-trifluoromethyl | |
| FC1=C(Br)C=C(C=N1)C(F)(F)F | |
| 243.99 | |
| 243.99 | |
| Liquid |
| 1.774 g/mL | |
| C6H2BrF4N | |
| MFCD08061304 | |
| UN1992 | |
| HKACHYLUXHBYMO-UHFFFAOYSA-N | |
| 3-bromo-2-fluoro-5-(trifluoromethyl)pyridine | |
| 23053237 | |
| 97% | |
| 3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only