missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-ethoxypyridine 97.0+%, TCI America™

Catalog No. B47381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47381G Supplier TCI America Supplier No. B47381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 57883-25-7
Color Yellow
Molecular Formula C7H8BrNO
Quantity 1 g
MDL Number MFCD00234309
Synonym 2-ethoxy-3-bromopyridine, pyridine, 3-bromo-2-ethoxy, 3-bromanyl-2-ethoxy-pyridine, 3-bromo-2-ethoxy-pyridine, pubchem6233, bromo-2 ethoxypyridine, 3-bromo-2-ethoxypyridine, pyridine,3-bromo-2-ethoxy
InChI Key KLQBFCAXXZMOLZ-UHFFFAOYSA-N
SMILES CCOC1=C(C=CC=N1)Br
IUPAC Name 3-bromo-2-ethoxypyridine
Molecular Weight (g/mol) 202.051
PubChem CID 10987308
Formula Weight 202.05
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-ethoxypyridine
Show More Show Less