missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-cyanopyridine 98.0+%, TCI America™
$140.54 - $408.13
Chemical Identifiers
| CAS | 55758-02-6 |
|---|---|
| Molecular Formula | C6H3BrN2 |
| Molecular Weight (g/mol) | 183.01 |
| MDL Number | MFCD02683288 |
| InChI Key | HCOPIUVJCIZALB-UHFFFAOYSA-N |
| Synonym | 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine |
| PubChem CID | 817694 |
| IUPAC Name | 3-bromopyridine-2-carbonitrile |
| SMILES | BrC1=CC=CN=C1C#N |
Chemical Identifiers
| 55758-02-6 | |
| 183.01 | |
| HCOPIUVJCIZALB-UHFFFAOYSA-N | |
| 817694 | |
| BrC1=CC=CN=C1C#N |
| C6H3BrN2 | |
| MFCD02683288 | |
| 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine | |
| 3-bromopyridine-2-carbonitrile |
Specifications
| 55758-02-6 | |
| White-Yellow | |
| C6H3BrN2 | |
| 1 g | |
| 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine | |
| BrC1=CC=CN=C1C#N | |
| 183.01 | |
| 183.01 | |
| Crystalline Powder |
| 99°C | |
| 120°C | |
| MFCD02683288 | |
| 3439 | |
| HCOPIUVJCIZALB-UHFFFAOYSA-N | |
| 3-bromopyridine-2-carbonitrile | |
| 817694 | |
| ≥98.0% (GC) | |
| 3-Bromo-2-cyanopyridine |
Safety and Handling
TSCA : No