missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-cyanopyridine 98.0+%, TCI America™

Catalog No. B43821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43821G 1 g
B43825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43821G Supplier TCI America Supplier No. B43821G

Chemical Identifiers

CAS 55758-02-6
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD02683288
InChI Key HCOPIUVJCIZALB-UHFFFAOYSA-N
Synonym 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine
PubChem CID 817694
IUPAC Name 3-bromopyridine-2-carbonitrile
SMILES BrC1=CC=CN=C1C#N

Specifications

Melting Point 99°C
Color White-Yellow
Boiling Point 120°C
Quantity 1 g
UN Number 3439
Formula Weight 183.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-cyanopyridine