missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH2763203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2763203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2763203 Supplier Thermo Scientific Chemicals Supplier No. H2763203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55758-02-6 | Molecular Formula: C6H3BrN2 | Molecular Weight: 183.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55758-02-6
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD02683288
InChI Key HCOPIUVJCIZALB-UHFFFAOYSA-N
Synonym 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine
PubChem CID 817694
IUPAC Name 3-bromopyridine-2-carbonitrile
SMILES BrC1=CC=CN=C1C#N

Specifications

Melting Point 96°C to 99°C
Boiling Point 120°C (4 mmHg)
Quantity 1 g
Formula Weight 183.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-cyanopyridine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only