missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-chlorothiophene 97.0+%, TCI America™
Supplier: TCI America B201325G
Specifications
| 3-Bromo-2-chlorothiophene | |
| 73°C | |
| MFCD00043883 | |
| 2-chloro-3-bromo thiophene, 2-chloro-3-bromothiophene, 3-bromo-2-chloro-thiophene, thiophene, 3-bromo-2-chloro, pubchem5259, acmc-1apw4, ksc491i4t, bidd:gt0537, 3-bromo-2-chlorothiophene, inchi=1/c4h2brcls/c5-3-1-2-7-4 3 6/h1-2 | |
| C1=CSC(=C1Br)Cl | |
| 197.474 | |
| 197.47 | |
| Liquid |
| 40032-73-3 | |
| C4H2BrClS | |
| 25 g | |
| KSHOQKKCPJELBV-UHFFFAOYSA-N | |
| 3-bromo-2-chlorothiophene | |
| 2724559 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 40032-73-3 | |
| 197.474 | |
| KSHOQKKCPJELBV-UHFFFAOYSA-N | |
| 2724559 | |
| C1=CSC(=C1Br)Cl |
| C4H2BrClS | |
| MFCD00043883 | |
| 2-chloro-3-bromo thiophene, 2-chloro-3-bromothiophene, 3-bromo-2-chloro-thiophene, thiophene, 3-bromo-2-chloro, pubchem5259, acmc-1apw4, ksc491i4t, bidd:gt0537, 3-bromo-2-chlorothiophene, inchi=1/c4h2brcls/c5-3-1-2-7-4 3 6/h1-2 | |
| 3-bromo-2-chlorothiophene |
Safety and Handling
TSCA : No