missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloropyridine 98.0+%, TCI America™

Catalog No. B23285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23285G 5 g
B232825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23285G Supplier TCI America Supplier No. B23285G
Only null left

Chemical Identifiers

CAS 52200-48-3
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD00234007
InChI Key HDYNIWBNWMFBDO-UHFFFAOYSA-N
Synonym 2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine
PubChem CID 693324
IUPAC Name 3-bromo-2-chloropyridine
SMILES C1=CC(=C(N=C1)Cl)Br

Specifications

Melting Point 57°C
Boiling Point 97°C
Quantity 5 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-chloropyridine