Learn More
3-Bromo-2-chloropyridine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 52200-48-3 |
|---|---|
| Molecular Formula | C5H3BrClN |
| Molecular Weight (g/mol) | 192.44 |
| InChI Key | HDYNIWBNWMFBDO-UHFFFAOYSA-N |
| Synonym | 2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine |
| PubChem CID | 693324 |
| IUPAC Name | 3-bromo-2-chloropyridine |
| SMILES | C1=CC(=C(N=C1)Cl)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC360650050
|
Thermo Scientific Chemicals
360650050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC360650250
|
Thermo Scientific Chemicals
360650250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 52200-48-3 | |
| 192.44 | |
| 2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine | |
| 3-bromo-2-chloropyridine |
| C5H3BrClN | |
| HDYNIWBNWMFBDO-UHFFFAOYSA-N | |
| 693324 | |
| C1=CC(=C(N=C1)Cl)Br |
Specifications
| 52200-48-3 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| HDYNIWBNWMFBDO-UHFFFAOYSA-N | |
| 3-bromo-2-chloropyridine | |
| 693324 | |
| 98% |
| 97.5 | |
| 97°C (1.3 kPa) | |
| 97.5% min. (GC) | |
| C5H3BrClN | |
| 2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine | |
| C1=CC(=C(N=C1)Cl)Br | |
| 192.44 | |
| 192.44 | |
| 3-Bromo-2-chloropyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
RUO â Research Use Only