missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chloropyridine-5-sulfonyl chloride, 96%

Catalog No. AAL1701503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1701503 1 g
AAL1701506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1701503 Supplier Thermo Scientific Chemicals Supplier No. L1701503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 216394-05-7 | C5H2BrCl2NO2S | 290.94 g/mol

3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Reacts with water. Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 216394-05-7
Molecular Formula C5H2BrCl2NO2S
Molecular Weight (g/mol) 290.94
MDL Number MFCD01318107
InChI Key TURGMVYIESHZBE-UHFFFAOYSA-N
Synonym 3-bromo-2-chloropyridine-5-sulfonyl chloride, 3-bromo-2-chloropyridine-5-sulphonyl chloride, 5-bromo-6-chloro-pyridine-3-sulfonyl chloride, 5-bromo-6-chloro-3-chlorosulphonyl pyridine, 5-bromo-6-chloro-3-pyridinesulfonyl chloride, 5-bromo-6-chloro-3-pyridine sulfonyl chloride, pubchem1289, 5-bromo-6-chloropyridine-3-sulfonylchloride, acmc-209xe4, 5-bromo-6-chloro 3-pyridyl chlorosulfone
PubChem CID 2793626
IUPAC Name 5-bromo-6-chloropyridine-3-sulfonyl chloride
SMILES ClC1=C(Br)C=C(C=N1)S(Cl)(=O)=O

Specifications

Melting Point 65°C to 70°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water. Soluble in dichloromethane.
Formula Weight 290.95
Percent Purity 96%
Chemical Name or Material 3-Bromo-2-chloropyridine-5-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only