missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,5-dichlorothiophene, 97%

Catalog No. p-7080871
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
AAA1060303 1 g
AAA1060306 5 g
AAA1060314 25 g
3 options

Catalog No. AAA1060303

Supplier: Thermo Scientific Chemicals A1060303

Only null left

CAS: 60404-18-4 | C4HBrCl2S | 231.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60404-18-4
Molecular Formula C4HBrCl2S
Molecular Weight (g/mol) 231.92
MDL Number MFCD00041440
InChI Key PBUHOXBSIQJRNO-UHFFFAOYSA-N
PubChem CID 521925
IUPAC Name 3-bromo-2,5-dichlorothiophene
SMILES ClC1=CC(Br)=C(Cl)S1

Specifications

Boiling Point 95°C to 97°C (14 mmHg)
Refractive Index 1.608
Quantity 1 g
Beilstein 1306272
Synonym thiophene,3-bromo-2,5-dichloro, zlchem 1326, acmc-209mjs, 2,5-dichloro bromo thiophene, 2,5-dichloro-3-bromothiophene, 3-bromo-2,5-dichloro thiophene, thiophene, 3-bromo-2,5-dichloro
Formula Weight 231.93
Percent Purity 97%
Chemical Name or Material 3-Bromo-2,5-dichlorothiophene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only