missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-2,2':5',2″-terthiophene 97.0+%, TCI America™

Catalog No. B40505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40505G 5 g
B40501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40505G Supplier TCI America Supplier No. B40505G

Chemical Identifiers

CAS 105125-00-6
Molecular Formula C12H7BrS3
Molecular Weight (g/mol) 327.272
InChI Key FGBHDLKMGUOJBA-UHFFFAOYSA-N
Synonym 3-Bromo-2,5-di(2-thienyl)thiophene
PubChem CID 184442
IUPAC Name 3-bromo-2,5-dithiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br

Specifications

Melting Point 40°C
Quantity 5 g
Formula Weight 327.27
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3′-Bromo-2,2′:5′,2′′-terthiophene