missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1H-indazole, 97%

Catalog No. AAH3777503
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3777503 1 g
AAH3777509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3777503 Supplier Thermo Scientific Chemicals Supplier No. H3777503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40598-94-5 | C7H5BrN2 | 197.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40598-94-5
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD00159926
InChI Key HTKXRTUKPXEALT-UHFFFAOYSA-N
PubChem CID 721568
IUPAC Name 3-bromo-2H-indazole
SMILES C1=CC2=C(NN=C2C=C1)Br

Specifications

Melting Point 148°C to 149°C
Quantity 1 g
UN Number UN2811
Synonym 3-bromoindazole, 3-bromo-1h-indazole, 1h-indazole, 3-bromo, bromoindazole, 3-brom-1h-indazol, 2h-indazole, 3-bromo, indazole, 3-bromo, zlchem 586, pubchem11861, 3-bromo-1 h-indazole
Formula Weight 197.04
Percent Purity 97%
Chemical Name or Material 3-Bromo-1H-indazole
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only