missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-trimethylsilyl-1-propyne, 98%

Catalog No. p-7035725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5345106 5 g
AAH5345114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5345106 Supplier Thermo Scientific Chemicals Supplier No. H5345106

CAS: 38002-45-8 | C6H11BrSi | 191.143 g/mol

3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

Solubility
Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 38002-45-8
Molecular Formula C6H11BrSi
Molecular Weight (g/mol) 191.143
MDL Number MFCD00134460
InChI Key GAPRPFRDVCCCHR-UHFFFAOYSA-N
Synonym 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r
PubChem CID 642589
IUPAC Name 3-bromoprop-1-ynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CCBr

Specifications

Boiling Point 44°C to 45°C (2mmHg)
Quantity 5 g
Linear Formula BrCH2C≡CSi(CH3)3
Beilstein 2037650
Flash Point 63°C (145°F)
Solubility Information Immiscible with water.
Formula Weight 191.14
Percent Purity 98%
Refractive Index 1.483
Density 1.17
Chemical Name or Material 3-Bromo-1-trimethylsilyl-1-propyne
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only