missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-propanol 93.0+%, TCI America™

Catalog No. p-7118583
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064125G 25 g
B0641250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B064125G Supplier TCI America Supplier No. B064125G
Only null left

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 138.99
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-propanol
EINECSNumber (2)-2002
RTECSNumber UA7385000
TSCA Yes