missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-propanol, 92%

Catalog No. AC403200050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403200050 5 g Glass bottle
AC403200250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403200050 Supplier Thermo Scientific Chemicals Supplier No. 403200050

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

CAS Min % 90.0
CAS Max % 100.0
Density 1.6000g/mL
Boiling Point 62.0°C (5.0 mmHg)
Flash Point 65°C
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4900
Linear Formula Br(CH2)3OH
Quantity 5 g
Beilstein 01, 356
Specific Gravity 1.6
Solubility Information Solubility in water: 167g/L water (20°C). Other solubilities: slightly soluble in ethanol and ether, soluble in most solvents
Formula Weight 138.99
Percent Purity 92%
Chemical Name or Material 3-Bromo-1-propanol, 92%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-986-6
RTECSNumber UA7385000
TSCA TSCA

RUO – Research Use Only