missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-benzofuran, 97%, Thermo Scientific™

Catalog No. p-4510324
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27510DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC27510DA Supplier Thermo Scientific Chemicals Supplier No. CC27510DA

Chemical Identifiers

CAS 59214-70-9
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
InChI Key ICJNAOJPUTYWNV-UHFFFAOYSA-N
Synonym 3-bromobenzofuran, 3-bromobenzo b furan, benzofuran, 3-bromo, 3-bromo-1-benzo b furan, 3-bromobenzofurane, 3-bromo-benzo b furan, 3-bromanyl-1-benzofuran, ksc269c4b
PubChem CID 640589
IUPAC Name 3-bromo-1-benzofuran
SMILES C1=CC=C2C(=C1)C(=CO2)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-1-benzofuran