Learn More
3-(Boc-aminomethyl)azetidine hydrochloride, 95%
CAS: 91188-15-7 | C9H18N2O2 | 186.26 g/mol
$159.04 - $266.56
Chemical Identifiers
| CAS | 91188-15-7 |
|---|---|
| Molecular Formula | C9H18N2O2 |
| Molecular Weight (g/mol) | 186.26 |
| MDL Number | MFCD01861760 |
| InChI Key | MOLUHRBHXXGWDP-UHFFFAOYSA-N |
| Synonym | 3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine |
| PubChem CID | 15346872 |
| SMILES | CC(C)(C)OC(=O)NCC1CNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH59816MD
|
Thermo Scientific Chemicals
H59816MD |
250 mg |
Each for $159.04
|
|
|||||
|
AAH59816ME
|
Thermo Scientific Chemicals
H59816ME |
500 mg |
Each for $266.56
|
|
|||||
Description
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 91188-15-7 | |
| 186.26 | |
| MOLUHRBHXXGWDP-UHFFFAOYSA-N | |
| 15346872 |
| C9H18N2O2 | |
| MFCD01861760 | |
| 3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine | |
| CC(C)(C)OC(=O)NCC1CNC1 |
Specifications
| 91188-15-7 | |
| MFCD01861760 | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)NCC1CNC1 | |
| 250 mg | |
| 15346872 | |
| 95% |
| C9H18N2O2 | |
| 3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine | |
| MOLUHRBHXXGWDP-UHFFFAOYSA-N | |
| tert-butyl N-(azetidin-3-ylmethyl)carbamate | |
| 186.26 | |
| 222.71 | |
| 3-(Boc-aminomethyl)azetidine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only