missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 99724-19-3 |
|---|---|
| Molecular Formula | C9H18N2O2 |
| Molecular Weight (g/mol) | 186.26 |
| InChI Key | DQQJBEAXSOOCPG-UHFFFAOYSA-N |
| Synonym | 3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine |
| PubChem CID | 2757234 |
| IUPAC Name | tert-butyl N-pyrrolidin-3-ylcarbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC452320010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 99724-19-3 | |
| 186.26 | |
| 3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine | |
| tert-butyl N-pyrrolidin-3-ylcarbamate |
| C9H18N2O2 | |
| DQQJBEAXSOOCPG-UHFFFAOYSA-N | |
| 2757234 | |
| CC(C)(C)OC(=O)NC1CCNC1 |
Specifications
| 99724-19-3 | |
| Glass Bottle | |
| 3-boc-aminopyrrolidine, tert-butyl pyrrolidin-3-ylcarbamate, 3-tert-butoxycarbonylamino pyrrolidine, 3-n-boc-aminopyrrolidine, 3-n-boc-amino pyrrolidine, 3-boc-amino pyrrolidine, tert-butyl n-pyrrolidin-3-yl carbamate, 3-tert-butoxycarbonylamino-pyrrolidine, 3-aminopyrrolidine, 3-boc protected, 3-tert-butoxycarbonyl amino pyrrolidine | |
| CC(C)(C)OC(=O)NC1CCNC1 | |
| 186.26 | |
| 186.26 | |
| 3-(BOC-amino)pyrrolidine |
| 96% min. (GC) | |
| C9H18N2O2 | |
| DQQJBEAXSOOCPG-UHFFFAOYSA-N | |
| tert-butyl N-pyrrolidin-3-ylcarbamate | |
| 2757234 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO – Research Use Only